Anti-IL1R2 Mouse Monoclonal Antibody (Fluorescein) [clone: 34141]

Supplier: R & D Systems
FAB663F
220017-950EA 404.92 USD
220017-950
Anti-IL1R2 Mouse Monoclonal Antibody (Fluorescein) [clone: 34141]
Antibodies
Type: Primary
Antigen: IL1R2
Clonality: Monoclonal
Clone: 34141
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR