Anti-CD44 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 740017]

Supplier: R & D Systems
FAB6577P
102883-316EA 438.18 USD
102883-316
Anti-CD44 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 740017]
Antibodies
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 740017
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR