Anti-ALCAM Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1592]

Supplier: R & D Systems
FAB6561P
220017-947EA 507.6 USD
220017-947
Anti-ALCAM Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1592]
Antibodies
Type: Primary
Antigen: ALCAM
Clonality: Monoclonal
Clone: 1592
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR