Anti-CD34 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 700011]

Supplier: R & D Systems
FAB6518G
102883-296EA 480.12 USD
102883-296
Anti-CD34 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 700011]
Antibodies
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 700011
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR