Anti-TACSTD2 Mouse Monoclonal Antibody (Fluorescein) [clone: 77220]

Supplier: R & D Systems
FAB650F
220017-945EA 404.92 USD
220017-945
Anti-TACSTD2 Mouse Monoclonal Antibody (Fluorescein) [clone: 77220]
Antibodies
Type: Primary
Antigen: TACSTD2
Clonality: Monoclonal
Clone: 77220
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR