Anti-PILRA Sheep Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R & D Systems
FAB6484P
102883-288EA 513.38 USD
102883-288
Anti-PILRA Sheep Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: PILRA
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR