Anti-PILRA Sheep Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
FAB6484A
102883-284EA 582.8 USD
102883-284
Anti-PILRA Sheep Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: PILRA
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR