Anti-CEACAM1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 723629]

Supplier: R & D Systems
FAB6480P
102883-282EA 438.18 USD
102883-282
Anti-CEACAM1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 723629]
Antibodies
Type: Primary
Antigen: CEACAM1
Clonality: Monoclonal
Clone: 723629
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR