Anti-CEACAM1 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 723629]

Supplier: R & D Systems
FAB6480G
102883-280EA 471.45 USD
102883-280
Anti-CEACAM1 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 723629]
Antibodies
Type: Primary
Antigen: CEACAM1
Clonality: Monoclonal
Clone: 723629
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR