Anti-VCAM1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 112734]

Supplier: R & D Systems
FAB6432P
220017-943EA 485.91 USD
220017-943
Anti-VCAM1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 112734]
Antibodies
Type: Primary
Antigen: VCAM1
Clonality: Monoclonal
Clone: 112734
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR