Anti-CDH2 Sheep Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
FAB6426G
102883-270EA 561.11 USD
102883-270
Anti-CDH2 Sheep Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen: CDH2
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 488
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR