Anti-CDH2 Sheep Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
FAB6426A
102883-268EA 582.8 USD
102883-268
Anti-CDH2 Sheep Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: CDH2
Clonality: Polyclonal
Clone: 0
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR