Anti-NPY1R Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 556153]

Supplier: R & D Systems
FAB6400A
102883-070EA 594.37 USD
102883-070
Anti-NPY1R Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 556153]
Antibodies
Type: Primary
Antigen: NPY1R
Clonality: Monoclonal
Clone: 556153
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR