Anti-EPHA2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 233720]

Supplier: R & D Systems
FAB639P
220017-941EA 501.82 USD
220017-941
Anti-EPHA2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 233720]
Antibodies
Type: Primary
Antigen: EPHA2
Clonality: Monoclonal
Clone: 233720
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR