Anti-EPHA2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 233720]

Supplier: R & D Systems
FAB639A
220017-940EA 561.11 USD
220017-940
Anti-EPHA2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 233720]
Antibodies
Type: Primary
Antigen: EPHA2
Clonality: Monoclonal
Clone: 233720
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR