Anti-LILRA2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 600007]

Supplier: R & D Systems
FAB6364A
102883-060EA 513.38 USD
102883-060
Anti-LILRA2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 600007]
Antibodies
Type: Primary
Antigen: LILRA2
Clonality: Monoclonal
Clone: 600007
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR