Anti-TNFRSF10D Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 104918]

Supplier: R & D Systems
FAB633P
220017-939EA 577.02 USD
220017-939
Anti-TNFRSF10D Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 104918]
Antibodies
Type: Primary
Antigen: TNFRSF10D
Clonality: Monoclonal
Clone: 104918
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR