Anti-TNFRSF10D Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 104918]

Supplier: R & D Systems
FAB633A
220017-937EA 608.83 USD
220017-937
Anti-TNFRSF10D Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 104918]
Antibodies
Type: Primary
Antigen: TNFRSF10D
Clonality: Monoclonal
Clone: 104918
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR