Anti-CD40 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 82111]

Supplier: R & D Systems
FAB6321A
220017-935EA 513.38 USD
220017-935
Anti-CD40 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 82111]
Antibodies
Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: 82111
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR