Anti-TNFRSF10C Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 90906]

Supplier: R & D Systems
FAB6302A
102883-238EA 594.37 USD
102883-238
Anti-TNFRSF10C Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 90906]
Antibodies
Type: Primary
Antigen: TNFRSF10C
Clonality: Monoclonal
Clone: 90906
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR