Anti-GPRC5D Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 571961]

Supplier: R & D Systems
FAB6300A
102883-236EA 639.2 USD
102883-236
Anti-GPRC5D Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 571961]
Antibodies
Type: Primary
Antigen Symbol: GPRC5D
Antigen Name: G protein-coupled receptor, class C, group 5, member D
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 571961
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR