Anti-LTBR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 71319]

Supplier: R & D Systems
FAB629A
220017-931EA 577.02 USD
220017-931
Anti-LTBR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 71319]
Antibodies
Type: Primary
Antigen: LTBR
Clonality: Monoclonal
Clone: 71319
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR