Anti-MUC1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 604804]

Supplier: R & D Systems
FAB6298G
102883-232EA 561.11 USD
102883-232
Anti-MUC1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 604804]
Antibodies
Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: 604804
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR