Anti-RAET1E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 709116]

Supplier: R & D Systems
FAB6285A
102883-220EA 614.62 USD
102883-220
Anti-RAET1E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 709116]
Antibodies
Type: Primary
Antigen: RAET1E
Clonality: Monoclonal
Clone: 709116
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR