Anti-TNFSF11 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 685857]

Supplier: R & D Systems
FAB6264A
102883-210EA 577.02 USD
102883-210
Anti-TNFSF11 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 685857]
Antibodies
Type: Primary
Antigen: TNFSF11
Clonality: Monoclonal
Clone: 685857
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR