Anti-TLR4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 610015]

Supplier: R & D Systems
FAB6248P
102883-208EA 543.75 USD
102883-208
Anti-TLR4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 610015]
Antibodies
Type: Primary
Antigen: TLR4
Clonality: Monoclonal
Clone: 610015
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR