Anti-TLR4 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 610015]

Supplier: R & D Systems
FAB6248N
102882-950EA 561.11 USD
102882-950
Anti-TLR4 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 610015]
Antibodies
Type: Primary
Antigen: TLR4
Clonality: Monoclonal
Clone: 610015
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR