Anti-MS4A3 Mouse Monoclonal Antibody (Fluorescein) [clone: 489433]

Supplier: R & D Systems
FAB6247F
102882-940EA 420.83 USD
102882-940
Anti-MS4A3 Mouse Monoclonal Antibody (Fluorescein) [clone: 489433]
Antibodies
Type: Primary
Antigen: MS4A3
Clonality: Monoclonal
Clone: 489433
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR