Anti-CD96 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 628211]

Supplier: R & D Systems
FAB6199A
220017-928EA 543.75 USD
220017-928
Anti-CD96 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 628211]
Antibodies
Type: Primary
Antigen: CD96
Clonality: Monoclonal
Clone: 628211
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR