Anti-CD40L Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 40804]

Supplier: R & D Systems
FAB617A
220017-918EA 513.38 USD
220017-918
Anti-CD40L Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 40804]
Antibodies
Type: Primary
Antigen: CD40L
Clonality: Monoclonal
Clone: 40804
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR