Anti-CD62E/CD62P Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: BBIG-E6 (13D5)]

Supplier: R & D Systems
FAB6169P
220017-917EA 389.02 USD
220017-917
Anti-CD62E/CD62P Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: BBIG-E6 (13D5)]
Antibodies
Type: Primary
Antigen: CD62E/CD62P
Clonality: Monoclonal
Clone: BBIG-E6 (13D5)
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR