Anti-IL13RA2 Goat Polyclonal Antibody (Fluorescein)

Supplier: R & D Systems
FAB614F
102882-912EA 420.83 USD
102882-912
Anti-IL13RA2 Goat Polyclonal Antibody (Fluorescein)
Antibodies
Type: Primary
Antigen: IL13RA2
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR