Anti-ENPP1 Sheep Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
FAB6136G
102883-204EA 566.89 USD
102883-204
Anti-ENPP1 Sheep Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen: ENPP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR