Anti-CSF2RA Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 698423]

Supplier: R & D Systems
FAB6130G
102883-196EA 543.75 USD
102883-196
Anti-CSF2RA Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 698423]
Antibodies
Type: Primary
Antigen: CSF2RA
Clonality: Monoclonal
Clone: 698423
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR