Anti-MHC class II (I-A/I-E) Rat Monoclonal Antibody (APC) [clone: M5/114.15.2]

Supplier: R & D Systems
FAB6118A
220017-912EA 594.37 USD
220017-912
Anti-MHC class II (I-A/I-E) Rat Monoclonal Antibody (APC) [clone: M5/114.15.2]
Antibodies
Type: Primary
Antigen Symbol: MHC class II (I-A/I-E)
Antigen Name:
Antigen Synonyms: H2-Ea
Clonality: Monoclonal
Clone: M5/114.15.2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR