Anti-CLEC9A Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 683409]

Supplier: R & D Systems
FAB6049N
102883-182EA 543.75 USD
102883-182
Anti-CLEC9A Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 683409]
Antibodies
Type: Primary
Antigen: CLEC9A
Clonality: Monoclonal
Clone: 683409
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR