Anti-CSF3R Sheep Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
FAB6039G
102883-176EA 513.38 USD
102883-176
Anti-CSF3R Sheep Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen: CSF3R
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Sheep
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR