Anti-CSF3R Sheep Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
FAB6039A
102883-174EA 549.54 USD
102883-174
Anti-CSF3R Sheep Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: CSF3R
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Sheep
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR