Anti-C3AR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 534625]

Supplier: R & D Systems
FAB5995A
220017-910EA 566.89 USD
220017-910
Anti-C3AR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 534625]
Antibodies
Type: Primary
Antigen: C3AR1
Clonality: Monoclonal
Clone: 534625
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR