Anti-NRP1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 761705]

Supplier: R & D Systems
FAB5994P
102883-172EA 480.12 USD
102883-172
Anti-NRP1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 761705]
Antibodies
Type: Primary
Antigen: NRP1
Clonality: Monoclonal
Clone: 761705
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR