Anti-NRP1 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 761705]

Supplier: R & D Systems
FAB5994N
102883-170EA 485.91 USD
102883-170
Anti-NRP1 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 761705]
Antibodies
Type: Primary
Antigen: NRP1
Clonality: Monoclonal
Clone: 761705
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR