Anti-NRP1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 761705]

Supplier: R & D Systems
FAB5994A
102883-166EA 561.11 USD
102883-166
Anti-NRP1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 761705]
Antibodies
Type: Primary
Antigen: NRP1
Clonality: Monoclonal
Clone: 761705
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR