Anti-LIFR Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 673602]

Supplier: R & D Systems
FAB5990P
102883-164EA 527.85 USD
102883-164
Anti-LIFR Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 673602]
Antibodies
Type: Primary
Antigen: LIFR
Clonality: Monoclonal
Clone: 673602
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR