Anti-CCR1 Rat Monoclonal Antibody (Fluorescein) [clone: 643854]

Supplier: R & D Systems
FAB5986F-100 FAB5986F-025
102883-156EA 438.18 USD
102883-156 102883-154
Anti-CCR1 Rat Monoclonal Antibody (Fluorescein) [clone: 643854]
Antibodies
Type: Primary
Antigen: CCR1
Clonality: Monoclonal
Clone: 643854
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR