Anti-TNFRSF17 Rat Monoclonal Antibody (Fluorescein) [clone: 161616]

Supplier: R & D Systems
FAB593F
220017-906EA 446.86 USD
220017-906
Anti-TNFRSF17 Rat Monoclonal Antibody (Fluorescein) [clone: 161616]
Antibodies
Type: Primary
Antigen: TNFRSF17
Clonality: Monoclonal
Clone: 161616
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR