Anti-MERTK Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 108928]

Supplier: R & D Systems
FAB5912A
102883-138EA 608.83 USD
102883-138
Anti-MERTK Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 108928]
Antibodies
Type: Primary
Antigen: MERTK
Clonality: Monoclonal
Clone: 108928
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR