Anti-CCR6 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 140706]

Supplier: R & D Systems
FAB590P-100 FAB590P-025
102883-136EA 527.85 USD
102883-136 102883-134
Anti-CCR6 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 140706]
Antibodies
Type: Primary
Antigen: CCR6
Clonality: Monoclonal
Clone: 140706
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR