Anti-CCR6 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 140706]

Supplier: R & D Systems
FAB590G-100 FAB590G-025
102883-128EA 527.85 USD
102883-128 102883-126
Anti-CCR6 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 140706]
Antibodies
Type: Primary
Antigen: CCR6
Clonality: Monoclonal
Clone: 140706
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR