Anti-TGFBR1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 141231]

Supplier: R & D Systems
FAB5871P-025 FAB5871P-100
102883-118EA 533.63 USD
102883-118 102883-116
Anti-TGFBR1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 141231]
Antibodies
Type: Primary
Antigen: TGFBR1
Clonality: Monoclonal
Clone: 141231
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR