Anti-TGFBR1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 141231]

Supplier: R & D Systems
FAB5871A-100 FAB5871A-025
102883-114EA 614.62 USD
102883-114 102883-112
Anti-TGFBR1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 141231]
Antibodies
Type: Primary
Antigen: TGFBR1
Clonality: Monoclonal
Clone: 141231
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR